Staphylococcus aureus Recombinant Protein A (from E. coli), Biotin
Supplier: Anaspec Inc
AS-72234
103010-690EA 328.85 USD
103010-690
Staphylococcus aureus Recombinant Protein A (from E. coli), Biotin
Proteins and Peptides
This is biotinylated Protein A conjugate suitable for asssay development, IHC, IF, immunoprecipitation or western blotting.
AnaSpec's recombinant Protein A consists of only IgG binding domains and was expressed using a recombinant bacterial expression system. Its apparent molecular weight is approximately 30,000 Da compared to 42,000 Da for the native Protein A that is found in Staphylococcus aureus.
Protein A conjugates are widely used as labeled reagents for a variety of experiments including immunoprecipitation, antibody detection and purification and assay development