AggreSure™ Human Beta-Amyloid (1-40)
Supplier: Anaspec Inc
AS-72215
103010-638EA
505.69
USD
103010-638
AggreSure™ Human Beta-Amyloid (1-40)
Proteins and Peptides
AnaSpec's AggreSure™ beta-Amyloid (1-40) peptide is pretreated and tested for aggregation using AnaSpec's SensoLyte® ThT Aβ40 Aggregation kit (Cat# AS-72213).
Sequence: DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVV
Molecular Weight: 4329.9 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...