AnaTag™ HiLyte™ Fluor 488 Protein Labeling Kit [3 labeling reactions (3×5 mg protein)], Anaspec
Supplier: Anaspec Inc
AS-72047
103010-354EA 697.15 USD
103010-354
AnaTag™ HiLyte™ Fluor 488 Protein Labeling Kit [3 labeling reactions (3×5 mg protein)], Anaspec
Protein Linking and Labeling Systems
HiLyte™ Fluor 488, SE is an excellent amine-reactive fluorescent labeling dye for generating protein conjugates. The spectrum of HiLyte™ Fluor488 is similar to fluorescein (FITC), resulting in an optimal match to filters designed for fluorescein.
HiLyte™ Fluor 488 is more photostable than FITC, providing researchers with additional time for capturing image. Its fluorescence can be detected at the emission wavelength of 523±10 nm when excited at 499±10 nm. The HiLyte™ Fluor 488-protein conjugates are very stable and suitable for immunofluorescent staining, in situ hybridization, flow cytometry and other biological applications. The kit has all essential components for performing the conjugation reaction and for purifying the HiLyte™ Fluor 488-protein conjugate. The entire process takes only half an hour. The kit is able to perform three conjugation reactions with each reaction labeling up to 5 mg IgG. Kit Size: 3 labeling reactions (3 x 5 mg protein)