Chicken OVA (323-339)
Supplier: Anaspec Inc
AS-27024
AS-72025
103000-572EA
219.23
USD
103000-572
103010-304
Chicken OVA (323-339)
Proteins and Peptides
An H-2b-restricted OVA class II epitope.
Sequence: ISQAVHAAHAEINEAGR
MW: 1773.9 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...