Human Recombinant MMP-2 (from E. coli)
Supplier: Anaspec Inc
AS-72005
103010-278EA 190 USD
103010-278
Human Recombinant MMP-2 (from E. coli)
Proteins and Peptides
Matrix metalloproteinases (MMPs) belong to a family of secreted or membrane-associated zinc endopeptidases capable of digesting extracellular matrix components. MMP-2 (72-kDa gelatinase-A) is involved in tumor development and metastasis. It is proposed as a therapeutic target for cancer.
Recombinant human MMP-2 was expressed as a pro-enzyme from its DNA sequence, transfected into CHO cells. The pro-MMP-2 can be fully activated by incubating with 1 mM APMA at 37°C for 20 min to 1 hr.Its activity can be measured in FRET-based enzymatic assays. 10-20 ng of enzyme is sufficient for FRET-based assay.