HiLyte™ Fluor 647 amine fluorescent dye
Supplier: Anaspec Inc
AS-81257
103010-194EA
407.77
USD
103010-194
HiLyte™ Fluor 647 amine fluorescent dye
HiLyte™ Fluor
HiLyte™ Fluor 647 amine is a carbonyl-reactive fluorescent labeling dye that generates the conjugates that are slightly red-shifted compared to those of Cy5 dyes, resulting in an optimal match to filters designed for Cy5 dye.
- Spectral Properties Abs/Em = 649/674 nm
- Solvent System water
- Storage -20°C desiccated and protected from light
Its conjugate may have better performance than Cy5 for fluorescence polarization-based assays.
Extinction Coefficient (M-1cm-1): 250,000
Fluorescence quantum yield: 0.33
Fluorescence Life Time (ns): 1.0
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...