Human Protease Activated Receptor 1
Supplier: Anaspec Inc
AS-65486
103010-016EA
146.15
USD
103010-016
Human Protease Activated Receptor 1
Proteins and Peptides
The peptide YFLLRNP is an antagonist to thrombin and SFLLRNP (thrombin receptor agonist peptide) in human platelets. YFLLRNP can induce partial activation of human platelets, visible by shape change; but it is unable to fully activate the platelets.
Sequence: YFLLRNP - OH
MW: 922.1 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...