Bovine P2 (53-78)
Supplier: Anaspec Inc
AS-65472
103009-986EA
454.54
USD
103009-986
Bovine P2 (53-78)
Proteins and Peptides
This peptide is derived from bovine peripheral myelin P2 protein amino acid residues 53-78. It is neuritogenic, inducing experimental autoimmune neuritis (EAN) in Lewis rats.
Sequence:TESPFKNTEISFKLGQEFEETTADNR
MW:3019.9 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...