[Lys(Me1)27/36]-Histone H3 (21-44)-GK,Biotin

Supplier: Anaspec Inc

AS-65446-1
103009-844EA 745.38 USD
103009-844
[Lys(Me1)27/36]-Histone H3 (21-44)-GK,Biotin
Proteins and Peptides

This peptide is Histone 3 amino acid residues 21 to 44. It is mono-methylated at Lys27 and Lys36 with a C-terminal G linker followed by a biotinylated lysine. Histone methylation plays an important role in the regulation of chromatin structure and function. Related Peptides: [Lys(Ac)27, Lys(Me1)36]-Histone H3 (21-44)-GK(Biotin), H3K27(Ac)K36(Me1), biotin-labeled, Cat# 65444 [Lys(Ac)27, Lys(Me2)36]-Histone H3 (21-44)-GK(Biotin), H3K27(Ac)K36(Me2), biotin-labeled, Cat# 65445
Sequence:ATKAAR-K(Me1)-SAPATGGV-K(Me1)-KPHRYRPG-GK(Biotin)
MW:2946.5 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR