Histone H3 (73-83)
Supplier: Anaspec Inc
AS-65437-1
103009-750EA
92.08
USD
103009-750
Histone H3 (73-83)
Proteins and Peptides
This peptide is based on amino acids 73 to 83 of histone H3. Lysine 79 of Histone H3 (H3K79) has been identified as a residue for acetylation or methylation.Related Peptides: [Lys(Ac)79]-Histone H3 (73-83), H3K79(Ac), Cat# 65438 [Lys(Me3)79]-Histone H3(73-83), H3K79(Me3), Cat# 65439[Lys(biotin)79]-Histone H3 (73-83), H3K79(biotin), Cat# 65440
Sequence:EIAQDFKTDLR
MW:1336.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...