[Arg(Me2s)3] - Histone H4 (1-21) - GGK,Biotin
Supplier: Anaspec Inc
AS-65424-1
103009-696EA
1346.08
USD
103009-696
[Arg(Me2s)3] - Histone H4 (1-21) - GGK,Biotin
Proteins and Peptides
This peptide is Histone H4 amino acid residues 1-21 with a C-terminal GG linker followed by a biotinylated lysine. Arginine 3 is dimethylated symmetrically.
Sequence:Ac-SG-R(me2s)-GKGGKGLGKGGAKRHRKV-GGK(biotin)
MW:2630.1 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...