Histone H3 (15-36)-GGK, Biotin
Supplier: Anaspec Inc
AS-65415-1
103009-532EA
629.92
USD
103009-532
Histone H3 (15-36)-GGK, Biotin
Proteins and Peptides
This peptide is Histone H3 amino acid residues 15-36 with a C-terminal GG linker followed by a biotinylated lysine.
Sequence:APRKQLATKAARKSAPATGGVK-GGK(biotin)
MW:2676.3 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...