[Lys(Ac)27]-Histone H3 (23-34)
Supplier: Anaspec Inc
AS-65387
103009-498EA
153.46
USD
103009-498
[Lys(Ac)27]-Histone H3 (23-34)
Proteins and Peptides
This is histone H3 (23-34) with acetylation at Lys27. Histone H3 acetylated at this residue is the least abundant isoform and is associated with histone deposition in replicating chromatin.
Sequence:KAAR-K(Ac)-SAPATGG
MW:1156.3 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...