Enhanced Green Fluorescent Protein
Supplier: Anaspec Inc
AS-65302
103009-386EA
157.85
USD
103009-386
Enhanced Green Fluorescent Protein
Proteins and Peptides
This peptide is H2-Kd-restricted enhanced green fluorescent protein (EGFP)-derived peptide (200-208) and represents a CD8 T cell epitope. Immunization with this peptide stimulates IFNg production, thus making this peptide a model tumor antigen for the experimental development of antigen-specific vaccines against cancer.
Sequence:HYLSTQSAL
MW:1019.1 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...