Histone H3 (5-23)
Supplier: Anaspec Inc
AS-65135
103009-012EA
206.08
USD
103009-012
Histone H3 (5-23)
Proteins and Peptides
This peptide represents amino acid residues 5-23 of histone H3 and can be used as a substrate for histone acetyl-transferase (HAT) assays.
Sequence:QTARKSTGGKAPRKQLASK
MW:2013.3 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...