Human [Pro18]-Beta-Amyloid (12-28)
Supplier: Anaspec Inc
AS-65113
103008-724EA
182.69
USD
103008-724
Human [Pro18]-Beta-Amyloid (12-28)
Proteins and Peptides
This peptide is derived from Aβ (12-28), which represents a binding site for apolipoprotein E (apoE). apoE is a genetically inherited risk factor for Alzheimer’s disease that promotes aggregation of toxic Aβ. Substitution of Val18 to proline competitively.Sequence: VHHQKLPFFAEDVGSNK
Molecular Weight: 1953.2 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...