Alpha-Gliadin (31-43)
Supplier: Anaspec Inc
AS-65106
103008-720EA
153.46
USD
103008-720
Alpha-Gliadin (31-43)
Proteins and Peptides
This peptide is derived from gliadin wheat protein residues 31-43. It elicits an innate immune response by upregulating expression of interleukin (IL)-15 and cyclooxygenase (COX)-2. This peptide also promotes expression of CD25 on monocytes and macrophages, expression of CD83 on dendritic cells, and p38 MAP kinase activation. Treatment with this peptide allows immunodominant epitopes (57-68 and 62-75) to induce T-cell activation and enterocyte apoptosis.
Sequence: LGQQQPFPPQQPY
MW: 1527.7 Da
% Peak area by HPLC: 95
Storage condition: -20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...