[Arg(Me1)3]-Histone H4(1-21)-GGK,Biotin
Supplier: Anaspec Inc
AS-64977-1
103008-636EA
660.62
USD
103008-636
[Arg(Me1)3]-Histone H4(1-21)-GGK,Biotin
Proteins and Peptides
This peptide is histone H4 amino acid residues 1 to 21. It is monomethylated at Arg3, with an acetylated N-terminus and C-terminal GG linker, followed by a biotinylated Lys. Methylation of histone H4 at Arg3 is an important step in transcriptional activation and allows for subsequent acetylation of H4 tails by p300. Provided at >90% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.'
Sequence:Ac-SG-R(Me1)-GKGGKGLGKGGAKRHRKV-GGK(Biotin)
MW:2616.1 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...