[Arg(Me2a)3]-Histone H4(1-21)-GGK,Biotin

Supplier: Anaspec Inc

AS-64976-1
103008-632EA 660.62 USD
103008-632
[Arg(Me2a)3]-Histone H4(1-21)-GGK,Biotin
Proteins and Peptides

This peptide is histone H4 amino acid residues 1 to 21. It is asymmetrically dimethylated at Arg3 with both methyl groups added to one nitrogen of the guanidinium group, and also contains a C-terminal GG linker, followed by a biotinylated Lys. Methylation of histone H4 at Arg3 is an important step in transcriptional activation, and allows for subsequent acetylation of H4 tails by p300. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:Ac-SG-R(Me2a)-GKGGKGLGKGGAKRHRKV-GGK(Biotin)
MW:2630.14 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR