Plasmodium Plasmepsin V FRET Substrate, DABCYL-EDANS
Supplier: Anaspec Inc
AS-64939
103008-598EA
363.92
USD
103008-598
Plasmodium Plasmepsin V FRET Substrate, DABCYL-EDANS
Proteins and Peptides
This FRET substrate peptide for Plasmepsin V (PMV) is derived from the conserved Plasmodium Export Element (PEXEL) motif of Histidine-Rich Protein II (HRPII). PMV is an ER aspartic protease that recognizes and cleaves the RXL sequence within the PEXEL motif of proteins exported by human malaria parasite Plasmodium falciparum, allowing them to translocate into host erythrocytes.
Sequence:Dabcyl-LNKRLLHETQ-EDANS
MW:1751.1 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...