L. monocytogenes LLO (91-99)
Supplier: Anaspec Inc
AS-64870
103008-564EA
150.54
USD
103008-564
L. monocytogenes LLO (91-99)
Proteins and Peptides
This peptide is the H2-Kd-restricted epitope of Listeriolysin O (LLO), consisting of amino acids 91 to 99. LLO is necessary for Listeria monocytogenes to escape the vacuoles of host cells and enter the cytoplasm during infection. This fragment has been shown to be a potential vaccine candidate against L. monocytogenes by eliciting a CTL response in vivo.
Sequence:GYKDGNEYI
MW:1058.1 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...