Lymphocytic Choreomeningitis Virus (LCMV) Glycoprotein 61-80
Supplier: Anaspec Inc
AS-64851
103008-550EA
216.31
USD
103008-550
Lymphocytic Choreomeningitis Virus (LCMV) Glycoprotein 61-80
Proteins and Peptides
This peptide is amino acids 61 to 80 fragment of the Lymphocytic Choriomeningitis Virus (LCMV) Glycoprotein (GP), an H-2Db restricted epitope capable of stimulating CD4+ T-cells.
Sequence:GLKGPDIYKGVYQFKSVEFD
MW:2290.6 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...