Histone H4 (1-23)-GSGSK, Biotin
Supplier: Anaspec Inc
AS-65002
103008-532EA
432.62
USD
103008-532
Histone H4 (1-23)-GSGSK, Biotin
Proteins and Peptides
This peptide consists of amino acids 1-23 of histone H4 attached at the C-terminal by a GSGS linker to biotinylated lysine. It is used as a substrate for histone acetyltransferase (HAT) assays. Biotinylation allows the peptide to be captured on streptavidin or agarose beads.
Sequence:SGRGKGGKGLGKGGAKRHRKVLR-GSGSK(Biotin)
MW:3003.6 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...