[Lys(Ac)5/8/12/16]-Histone H4 (1-21)-GGK,Biotin

Supplier: Anaspec Inc

AS-64989-1
103008-528EA 691.31 USD
103008-528
[Lys(Ac)5/8/12/16]-Histone H4 (1-21)-GGK,Biotin
Proteins and Peptides

This peptide is histone H4 (1-21), acetylated at lysine 5, 8, 12, and 16. It contains a C-terminal GG linker, followed by a biotinylated Lys. In general, histone H4 acetylation is associated with increased DNA replication during mitosis, although different combinations of acetylation at specific sites are selective for specific processes. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:SGRG-K(Ac)-GG-K(Ac)-GLG-K(Ac)-GGA-K(Ac)-RHRKV-GGK(Biotin)
MW:2728.2 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR