Human [Lys(Ac)382]-p53 (361-393), Biotin, AnaSpec
Supplier: Anaspec Inc
AS-65045-1
103008-518EA
691.31
USD
103008-518
Human [Lys(Ac)382]-p53 (361-393), Biotin, AnaSpec
Proteins and Peptides
This peptide is derived from amino acid residues 372-389 of human p53 tumor suppressor protein and is acetylated at Lys382. It is biotinylated at its N-terminus through a 6-carbon LC linker. Activated p53 functions to induce cell cycle arrest and apoptosis in response to signals such as DNA damage. Acetylation of p53 reduces ubiquitination by MDM2 and increases p53 half-life in vivo.
Sequence:Biotin-LC-KKGQSTSRHK-K(Ac)-LMFKTEG
MW:2473 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...