[Lys(Ac)27]-Histone H3 (21-43)-GGK(Biotin),Biotin
Supplier: Anaspec Inc
AS-64846-1
103008-492EA
507.15
USD
103008-492
[Lys(Ac)27]-Histone H3 (21-43)-GGK(Biotin),Biotin
Proteins and Peptides
This peptide is Histone H3 (21-43) acetylated at lysine 27 with a C-terminal GG linker followed by a biotinylated lysine. The acetylation of histone H3 at lysine 27 is associated with many active mammalian genes. In Arabidopsis thaliana, the acetylated histone at lysine 27 is nontransposable element gene-specific. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:ATKAAR-K(Ac)-SAPATGGVKKPHRYRP-GGK(Biotin)
MW:2959.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...