Pig Protegrine-1 (PG-1)
Supplier: Anaspec Inc
AS-64819-05
103008-470EA 587.54 USD
103008-470
Pig Protegrine-1 (PG-1)
Proteins and Peptides
This peptide is Protegrin-1 (PG-1) with a modified C-terminal amide. PG-1 is an 18-amino-acid beta-hairpin antimicrobial peptide found in porcine leukocytes and belongs to the cathelicidin family. PG-1 exhibits broad-spectrum activity unaffected by extracellular NaCl concentrations and is capable of inactivating numerous bacterial strains, Candida albicans, and some enveloped viruses. The efficacy is strongly dependent upon the existence of two disulfide bonds that stabilize the beta-sheet structure.