Human Endothelin 1, HiLyte™ Fluor 488
Supplier: Anaspec Inc
AS-64817-025
103008-468EA
767.31
USD
103008-468
Human Endothelin 1, HiLyte™ Fluor 488
Proteins and Peptides
This is a fluorescent (HiLyte™ Fluor 488)-labeled Endothelin 1 (ET-1) peptide, Abs/Em=503/528 nm.
Sequence:HiLyte™ Fluor 488-CSCSSLMDKECVYFCHLDIIW (Disulfide Bridge: 1-15 and 3-11)
MW:2848.4 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...