Chicken OVA (329-337)
Supplier: Anaspec Inc
AS-64777
103008-446EA
153.46
USD
103008-446
Chicken OVA (329-337)
Proteins and Peptides
This sequence is OVA (ovalbumin) peptide residues 257 to 280, also known as OT-II (OVA-specific T-cell receptor) peptide, the core H2b-restricted class II MHC epitope.
Sequence: AAHAEINEA
MW: 925 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...