Ryanodine receptor
Supplier: Anaspec Inc
AS-64769
103008-442EA
532
USD
103008-442
Ryanodine receptor
Proteins and Peptides
This is amino acids 2460 to 2495 fragment of cardiac ryanodine receptor (RyR2). RyR2 controls calcium release from the sarcoplasmic reticulum, which begins muscle contraction. Mutated RyR2 is associated to ventricular tachycardia (VT) and sudden death
Sequence:GFCPDHKAAMVLFLDRVYGIEVQDFLLHLLEVGFLP
MW:4103.9 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...