Chicken OVA (323-339), FITC (Fluorescein Isothiocyanate)
Supplier: Anaspec Inc
AS-64766
103008-438EA
451.62
USD
103008-438
Chicken OVA (323-339), FITC (Fluorescein Isothiocyanate)
Proteins and Peptides
This fluorescent (FITC)-labeled OVA (323 - 339) Peptide is an H-2b-restricted OVA class II epitope (Abs/Em = 493/522 nm).
Sequence: FITC-LC-ISQAVHAAHAEINEAGR
MW: 2276.5 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...