Smac/Diablo Peptide [AVPIAQKSE]
Supplier: Anaspec Inc
AS-64758
103008-434EA
415.08
USD
103008-434
Smac/Diablo Peptide [AVPIAQKSE]
Proteins and Peptides
This is a 5-FAM-labeled Smac/Diablo Peptide (Abs/Em=492/518 nm), which serves as a Livin inhibitor. Livin prevents apoptosis and sensitizes Livin-expressing cells to chemotherapy. This peptide has the potential to be used as a therapeutic agent in cancer treatment.
Sequence:AVPIAQKSEK-K(5-FAM)-NH2
MW:1555.8 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...