Vaccinia Virus B8R (20-27)
Supplier: Anaspec Inc
AS-64688
103008-426EA
138.85
USD
103008-426
Vaccinia Virus B8R (20-27)
Proteins and Peptides
This is amino acids 20 to 27 fragment of B8R, a vaccinia virus (VV) gene that encodes a secreted protein related to gamma interferon receptor (IFN-g). B8R binding to IFN-g neutralizes its antiviral activity.
Sequence:TSYKFESV
MW:960.1 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...