[Lys(Me3)20]-Histone H4 (1-23)-GGK,Biotin
Supplier: Anaspec Inc
AS-64625-1
103008-342EA
629.92
USD
103008-342
[Lys(Me3)20]-Histone H4 (1-23)-GGK,Biotin
Proteins and Peptides
This peptide is Histone H4 (1-23). It is trimethylated at lysine 20 with a C-terminal GG linker, followed by a biotinylated lysine. Methylation of histone H4 at lysine 20 has a critical role in gene expression, cell cycle progression, and DNA repair. The trimethylation of histone H4 at lysine 20 maintains heterochromatin structures. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:SGRGKGGKGLGKGGAKRHR-K(Me3)-VLR-GGK(Biotin)-NH2
MW:2870.4 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...