[Lys(Me1)20]-Histone H4 (1-23)-GGK,Biotin
Supplier: Anaspec Inc
AS-64623-1
103008-334EA
629.92
USD
103008-334
[Lys(Me1)20]-Histone H4 (1-23)-GGK,Biotin
Proteins and Peptides
This peptide is Histone H4 (1-23). It is monomethylated at lysine 20 with a C-terminal GG linker, followed by a biotinylated lysine. The monomethylation of histone H4 at lysine 20 is proposed to be a crucial mark for chromosomal memory. It is also very active throughout the cell cycle and is abundant during S phase. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:SGRGKGGKGLGKGGAKRHR-K(Me1)-VLR-GGK(Biotin)-NH2
MW:2842.4 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...