4N1K, AnaSpec
Supplier: Anaspec Inc
AS-64591
103008-256EA
122.77
USD
103008-256
4N1K, AnaSpec
Proteins and Peptides
4N1K is a cell-binding domain adhesive peptide. It has been identified as an IAP agonist. Integrin-associated protein (IAP) is important in host defense where it is required for integrin-dependent functions of polymorphonuclear leukocytes. IAP also appears to be important in modulating integrin function in other cells and in signal transduction upon ligand binding by certain integrins with which it associates.
Sequence:KRFYVVMWKK
MW:1384.8 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...