Lymphocytic Choreomeningitis Virus (LCMV) Glycoprotein 61
Supplier: Anaspec Inc
AS-64560
103008-236EA
239.69
USD
103008-236
Lymphocytic Choreomeningitis Virus (LCMV) Glycoprotein 61
Proteins and Peptides
An H-2Db restricted epitope, this peptide is amino acids 61 to 80 fragment of the lymphocytic choriomeningitis virus (LCMV) pre-glycoprotein polyprotein GP complex. LCMV has been routinely used for the study of adaptive immune responses to viral infection.
Sequence:GLNGPDIYKGVYQFKSVEFD
MW:2276.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...