[Lys(Me3)79]-Histone H3 (69-89)-K,Biotin
Supplier: Anaspec Inc
AS-64545-025
103008-230EA
251.38
USD
103008-230
[Lys(Me3)79]-Histone H3 (69-89)-K,Biotin
Proteins and Peptides
This peptide is Histone H3 amino acid residues 69 to 90. It is trimethylated at lysine 79 with a biotinylated lysine added to the C-terminus. The trimethylation of histone H3 at lysine 79 is found in the pericentromeric heterochromatin regions where active genes are not present. Provided at >90% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:RLVREIAQDF-K(Me3)-TDLRFQSSAV-K(Biotin)
MW:2876.3 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...