Prostatic Acid Phosphatase (248-286)
Supplier: Anaspec Inc
AS-64518
103008-214EA
621.15
USD
103008-214
Prostatic Acid Phosphatase (248-286)
Proteins and Peptides
Prostatic Acid Phosphatase (248-286), PAP (248-286) peptide is a semen-derived enhancer of viral infection (SEVI) factor found in semen. This peptide greatly increases HIV infection through enhanced virion attachment to target cells.
Sequence:GIHKQKEKSRLQGGVLVNEILNHMKRATQIPSYKKLIMY
MW:4551.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...