[Lys(Me1)4]-Histone H3 (1-21)
Supplier: Anaspec Inc
AS-64459
103008-186EA
558.31
USD
103008-186
[Lys(Me1)4]-Histone H3 (1-21)
Proteins and Peptides
This is a monomethylated lysine at position 4 of the 1-21 amino acid histone H3 peptide. This form of histone methylation is characterized as being enriched around transcription start sites. It’s chromatin mark was also closely correlated to cell-type specific expression of putative gene targets of enhancers. It was also noted from a genome wide study that most potential regulatory elements were enriched only by the mono-methylated version of this histone 3.
Sequence:ART-K(Me1)-QTARKSTGGKAPRKQLA
MW:2268.6 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...