Human Amylin,Biotin
Supplier: Anaspec Inc
AS-64451-05
103008-174EA
613.85
USD
103008-174
Human Amylin,Biotin
Proteins and Peptides
This amidated amylin (1-37) peptide has a biotin conjugated on the N-terminus. Amylin (1-37) or IAPP (Islet Amyloid Polypeptide Precursor) is a major constituent of protein deposits identified in the Islets of Langerhans of patients with noninsulin-dependent diabetes mellitus.
Sequence: Biotin - KCNTATCATQRLANFLVHSSNNFGAILSSTNVGSNTY - NH2 (disulfide bridge: 2 - 7)
MW: 4129.7 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...