[Lys(Me2)36]-Histone H3 (21-44)-GK,Biotin
Supplier: Anaspec Inc
AS-64442-025
AS-64442-1
103008-166EA
225.08
USD
103008-166
103008-168
[Lys(Me2)36]-Histone H3 (21-44)-GK,Biotin
Proteins and Peptides
This is a Histone 3 lysine 36 dimethylated peptide. This peptide has been shown to recruit histone deacetylase complex with nucleosomes and repress transcription. In addition, owing to its epigenetic repressive mark, it allows demethylation by a Jumonji C-domain family member, JMJD5 that functions as a transcription activator by inhibiting HDAC, and regulates cell cycle proliferation.
Sequence:ATKAARKSAPATGGV-K(Me2)-KPHRYRPG-GK(Biotin)
MW:2945.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...