Mant-GTP 5 mM in Tris/EDTA (TE) buffer (pH 7.6)

Supplier: Anaspec Inc

Synonyms: 2'-/3'-O-(N'-Methylanthraniloyl)guanosine-5'-O-triphosphate, trisodium salt

AS-64609
103008-100EA 600.69 USD
103008-100
Mant-GTP 5 mM in Tris/EDTA (TE) buffer (pH 7.6)
Mant-GTP

Mant-GTP is a fluorescent analog of GTP with Ex/Em = 355/448 nm. The fluorescence quantum yield of Mant fluorophore is 0.2 in water which increases significantly in nonpolar solvents or upon binding to most proteins. This highly environmental sensitive fluorescence of Mant makes Mant-GTP useful for directly detecting the nucleotide-protein interactions.


  • Spectral properties: Abs/Em = 355/448 nm
  • Storage Temperature: -20°C
  • Shipping condition: Blue ice

Formula: C₁₈H₂₀N₆Na₃O₁₅P₃
MW: 722.28 g/mol
Storage Temperature: Freezer
CAS Number: 148821-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR