[Lys(Me3)36]-Histone H3 (31-41)
Supplier: Anaspec Inc
AS-64381-1
103008-040EA
330.31
USD
103008-040
[Lys(Me3)36]-Histone H3 (31-41)
Proteins and Peptides
This peptide is Histone H3 amino acid residues 31 to 41 tri-methylated at Lys-36.
Sequence:STGGV-K(Me3)-KPHRY
MW:1271.4 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...