[Lys(Ac)9]-Histone H3 (1-21)
Supplier: Anaspec Inc
AS-64191
103007-962EA
380
USD
103007-962
[Lys(Ac)9]-Histone H3 (1-21)
Proteins and Peptides
This peptide sequence is found in residues 1 to 21 of the histone H3K9(Ac) . H3K9(Ac) is highly localized to the 5’ region of transcription start sites in human genes. Localization of H3K9(Ac) to the transcriptionally active 5’ region of human genes suggests this peptide is essential for transcription initiation and elongation.
Sequence:ARTKQTAR-K(Ac)-STGGKAPRKQLA
MW:2296.7 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...