Human Beta-Amyloid (1-42), HiLyte Fluor® 647
Supplier: Anaspec Inc
This beta-amyloid (1-42) peptide is labeled on the N-terminus with HiLyte™ Fluor 647, Abs/Em = 649/674 nm.
Sequence: HiLyte™ Fluor 647-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
MW: 5449.4 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Citations:
Esbjörner, EK. et al. (2014). Direct Observations of Amyloid β Self-Assembly in Live Cells Provide Insights into Differences in the Kinetics of Aβ (1–40) and Aβ (1–42) Aggregation. Chem Biol 21, 732. doi: 10.1016/j.chembiol.2014.03.014.
Narayan, P. et al. (2014). Rare Individual Amyloid-β Oligomers Act on Astrocytes to Initiate Neuronal Damage. Biochem 53, 2442. doi: 10.1021/bi401606f.
Quinn, SD. et al. (2014). Real-time probing of β-amyloid self-assembly and inhibition using fluorescence self-quenching between neighbouring dyes. Mol BioSyst 10, 34. doi: 10.1039/C3MB70272C.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...