Srctide,Biotin
Supplier: Anaspec Inc
AS-64108
103007-842EA
219.23
USD
103007-842
Srctide,Biotin
Proteins and Peptides
This peptide substrate for many protein kinases, such as Blk, BTK, cKit, EPHA1, EPHB2, EPHB3, ERBB4, FAK, Flt3, IGF-1R, ITK, Lck, MET, MUSK, Ret, Src, TIE2, TrkB, VEGF-R1 (Flt-1) and VEGF-R2 (KDR) is biotinylated on the N-terminus.
Sequence:Biotin-GEEPLYWSFPAKKK-NH2
MW:1905.3 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...