Srctide, FAM (Carboxyfluorescein)
Supplier: Anaspec Inc
AS-64106
103007-840EA
454.54
USD
103007-840
Srctide, FAM (Carboxyfluorescein)
Proteins and Peptides
This peptide is Srctide with an N-terminal 5-FAM (Ex/Em=494/521 nm) label. Srctide is a substrate for many protein kinases, such as Blk, BTK, cKit, EPHA1, EPHB2, EPHB3, ERBB4, FAK, Flt3, IGF-1R, ITK, Lck, MET, MUSK, Ret, Src, TIE2, TrkB, VEGF-R1 (Flt-1) and VEGF-R2 (KDR).
Sequence:5-FAM-GEEPLYWSFPAKKK-NH2
MW:2037.3 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...