Bcl-2 Binding Peptide
Supplier: Anaspec Inc
AS-64082
103007-830EA
371.23
USD
103007-830
Bcl-2 Binding Peptide
Proteins and Peptides
This is a bcl-2 binding peptide. This peptide is derived from the BH3 domain (a death domain) of Bad, amino acid residues 140 to 165.
Sequence:LWAAQRYGRELRRMSDEFEGSFKGL
MW:3003.4 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...