(Arg)9,Biotin
Supplier: Anaspec Inc
AS-64078
103007-820EA
206.08
USD
103007-820
(Arg)9,Biotin
Proteins and Peptides
(Arg)9 is a cell-permeable peptide used for drug delivery.It can traverse the plasma membrane of eukaryotic cells. This peptide sequence with nine arginines contains a biotin group attached to the epsilon amino group of lysine at the N-terminus.
Sequence:Biotin-LC-RRRRRRRRR-NH2
MW:1762.2 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...